Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0270L
101294-868EA 171 USD
101294-868
Taq 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq 2X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI and stabilizers
It is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.