Syringe Filters, PerkinElmer
Supplier: PerkinElmer
02542884 02542783 02542880 02542881 02542882 02542885 02542886 02542900 02542901 02542902 02542903 02542905 02542907 02542909 02542913 02542915 02542922 02542925
97019-098EA 545.14 USD
97019-098 97019-089 97019-090 97019-092 97019-094 97019-096 97019-100 97019-102 10129-658 10129-660 10129-662 10129-664 10129-666 10129-668 10129-670 10129-672 10129-674 10129-676 10129-678 10129-680 10129-682
Syringe Filters, PerkinElmer
Filters Syringe Filters
These 17 mm syringe filters are ideal for coarse/crude samples or higher detection limit applications such as food, beverage, or environmental analysis.
  • Ideal for routine QA/QC analysis
Nylon Syringe Filters: Ideal for aqueous (non-acidic) or organic sample preparation and HPLC, GC, or dissolution sample analysis.
PTFE (Hydrophobic) Syringe Filters: Versatile filters for use with aggressive organic solvent-based solutions, and especially ideal for HPLC sample preparation. Hydrophobic PTFE syringe filters have broad chemical compatibility and highMore Product Information