Syringes with BD Luer-Lok® Tip, Autosampler, PerkinElmer
Supplier: PerkinElmer
02542891
10129-657PK 407.25 USD
10129-657
Syringes with BD Luer-Lok® Tip, Autosampler, PerkinElmer
Syringes Syringes for Chromatography
Syringe with BD Luer-Lok® Tip is a autosampler syringe useful in 200 series autosampler.
  • Increased secure connection
  • Tapered plunger rod
  • Positive plunger rod stop
  • Bold graduation markings
BD™ syringes feature a clear barrel with bold scale markings, tapered plunger rod for ease of aspiration, positive plunger rod stop, and an added BD Luer-Lok™ thread for increased secure connection.