E.Z.N.A.® Micro RNA Kits, Omega Bio-Tek
Supplier: OMEGA BIO-TEK
R7034-01
10129-298EA 361.57 USD
10129-298
E.Z.N.A.® Micro RNA Kits, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A
The system uses RNA-Solv to efficiently lyse cells and protect RNA from degradation. Following a chloroform separation step, RNA is applied to a HiBind RNA column where large RNA binds. The flow through is retained and applied to a HiBind Micro RNA column to bind small RNA.
Micro RNA Kit combines the reversible binding properties of HiBind® matrix, a new silica-based material, with the highly efficient lysis ability of RNA-Solv® to extract micro and large RNA from a wide variety of starting materials. Micro and large RNA are separated to allow for quantification into downstream applications.