Radleys Reactor-Ready Duo Lab Reactor
Supplier: Heidolph NA, LLC
015970100
10129-172EA 29833.3 USD
10129-172
Radleys Reactor-Ready Duo Lab Reactor
Laboratory Reactor Systems
Radleys reactor-ready duo lab reactor shares the same unique features as Radleys reactor-ready lab reactor, except that it supports two separate jacketed glass reaction vessels
Rapid exchange of both vessels independently, with quick-release vessel clamp and hose couplings. Choice of manifold kits allow two vessels to run from a single thermo regulator or two separte thermo regulators.
The system accepts two overhead stirrers which can be operated and moved independently. Robust, heavy-duty framework features solid stainless steel support rods. Two separate quick-release clamps support each of the vessels independently. Large drip tray will retain vessel contents.
For parallel synthesis or reaction optimization, use similar or different size vessels and vary stirring speed, stirrer shape, and temperature between vessels. For two-stage reactions, transfer reactant from one vessel to the other using a vacuum or a pump. For single reaction vessel, use the second vessel as either a receiving or feed vessel (where reagent can be pre-heated or pre-cooled prior to addition).
The system can be configured to operate with a single thermo regulator controling the temperature of both vessels simultaneously, or with two thermo regulators controling the temperature of each vessel independently.
Ordering information: Vessel kit and overhead stirrer sold separately. Manifold and hose kits must be ordered separately.