Blade and Half-Moon Impeller
Supplier: Heidolph NA, LLC
036304390
10129-160EA 202.1 USD
10129-160
Blade and Half-Moon Impeller
Stirrers Stirring Blades
These impellers are recommended for applications which require average speed.
Designed for mixing tasks with little or average viscosity. The blade is constructed from stainless steel, AISI 304.