Cryopreservation Media, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
Cryopreservation Medium is a ready-to-use medium for the preservation of cells as seed stock or for future investigative study. Cryopreservation allows you to preserve and protect your cell line stock(s) from over culturing that can lead to unwanted changes within cellular properties or lead to senescence.
- Ready-to-use
- Optimizes cell growth and cell viability after thawing
- Reduces potential for transmission of infectious diseases
- Contains Fetal Bovine Serum & DMSO
The FBS in the Cryopreservation Medium improves the post-freezing recovery and survival after cryopreservation, while DMSO preserves the cells during freeze. Cryopreservation Medium with Fetal Bovine Serum (FBS) and Calf Serum is a complete cryopreservation medium with 10% DMSO.
Sterile Filtered
Performance Tested
Harmonization Code: 3821.00.0000
ECCN: EAR99
Contents: DMEM, 1.289 g/L, phenol red, 0.01 g/L, fetal bovine serum, L-glutamine, 0.584 g/L, DMSO, 100 ml/L
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...