Glutamine XL 200 mMr, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
118-288-061
10128-832EA 63.96 USD
10128-832
Glutamine XL 200 mMr, Quality Biological
Cell Culture Supplements and Additives
Glutamine XL is useful as a cell culture media supplement
  • With stable L-Alanyl-Glutamine
Standard, unstabilized solutions of L-glutamine spontaneously break down over time to release cytotoxic amines. Glutamine XL is a stabilized solution that prevents degradation and cytotoxicity, with the benefit of a long shelf life.
The incorporated L-alanine protects the unstable alpha amino acid group of L-glutamine. Aminopeptidase within the cells cleaves the dipeptide, gradually releasing both L-glutamine and L- alanine for use.
Recommendation
Dilute Glutamine XL the same way as L-glutamine in the appropriate cell culture medium. Typically, L-glutamine is diluted to concentration of 1-4 mM. Protect from light.
Sterile Filtered
Endotoxin and Mycoplasma Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.
Caution: Research Use Only