ENZ-Free, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
118-094-721
10128-796EA 110.85 USD
10128-796
ENZ-Free, Quality Biological
Cell Separation Media
Enz-Free Solution is a proprietary, non-enzymatic solution specially formulated for the non-destructive detachment of adherent cells from culture vessel surfaces. Enz-Free is a gentle dissociation of mammalian cells for studies requiring intact cell surface proteins such as ligand binding, flow cytometry, and immunohistochemistry.
  • Recommended for detaching adherent cells growing in serum free conditions
Enz-Free works well with lightly adherent cell lines such as HeLa and NIH 3T3, but is not recommended for routine passage of strongly adherent cells. Enz-Free is a membrane-filtered, isotonic, and enzyme-free solution of salts, chelating agents, and cell-conditioning agents in Dulbecco’s Phosphate Buffered Saline (DPBS) without calcium and magnesium.
Enz-Free is useful for serum-containing cell culture systems, serum-free cell culture systems, and protein-free cell culture systems.
Recommendation
Use ENZ-Free with accessory item QBSF 60 (Cat No. 10128-236) serum free cell culture medium.
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months
Enz-Free is a gentle dissociation of mammalian cells for studies requiring intact cell surface proteins such as ligand binding, flow cytometry, and immunohistochemistry.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Delivery information: Contents: Chelating, stabilizing, and cell-conditioning agents
Caution: Research Use Only