Sodium chloride 0.85% in aqueous solution, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

114-180-101
10128-752EA 40.53 USD
10128-752
Sodium chloride 0.85% in aqueous solution, sterile filtered
Sodium chloride

Sodium Chloride, 0.85% is a saline solution at 0.85% prepared with water used for cell culture. The sodium chloride is a source of chlorine and sodium necessary to the production of components like sodium carbonate, sodium hydroxide or chlorate ions.


  • Made with cell culture grade water


Sodium Chloride, 0.85% is useful in the following applications: flushing and cleansing, preparation of plasmidic DNA (removing of small fragments of nucleic acids), realisation of phosphate buffer or SSC buffer (sodium chloride – sodium citrate), precipitation of the DNA of samples containing SDS, chemical sequencing of DNA, and protein crystallisation.

Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR