Tris Glycine Transfer Buffer 10X (Transblotting Buffer 10X), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-087-131
10128-706EA 95.81 USD
10128-706
Tris Glycine Transfer Buffer 10X (Transblotting Buffer 10X), Quality Biological
Buffers Electrophoresis Buffers Transfer Buffers
TRIS Glycine Transfer Buffer, 10X (Transblot Buffer, 10X).
Tris Glycine Transfer Buffer, at a 1X concentration with the addition of 20% methanol, is used for electrophoretic transfer of proteins from denaturing polyacrylamide gel to a solid membrane – i.e. Western blotting or Tris-glycine gel electrophoresis.
Tris Glycine Transfer Buffer (10X) is prepared from molecular biology grade Tris base [tris(hydroxymethyl)- aminomethane] and glycine using Quality Biological’s Molecular Biology Grade (MBG) Water. The final product is sterile filtered using a 0.2 µm filter.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.