YPAD Agar, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
342-310-219
10128-678CS 667.42 USD
10128-678
YPAD Agar, Quality Biological
Media for Microbiology Prepared Media for Microbiology
YPAD with Adenine Sulfate Agar is used to inhibit reversion of the ade1 and ade2 mutations.YPAD with Adenine Sulfate Agar inhibits red coloration in ade strains.
YPAD with Adenine Sulfate Agar is useful for detecting carcinogens with this yeast assay and preparing yeast stock for long-term storage.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only