Fischer’s Medium with L-Glutaminer, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
112-032-101
10128-634EA 48.13 USD
10128-634
Fischer’s Medium with L-Glutaminer, Quality Biological
Cell Culture Media
Fischer’s medium was originally developed by Glen Fischer at the National Cancer Institute (Bethesda, MD), for serial propagation of cells from leukemic mice. Fischer’s medium supports clonal growth of cells – particularly lymphoblasts – from primary explants or cells already in culture.
  • With L-glutamine and Phenol Red
Fischer’s medium also supports mammalian cell culture and murine hematopoietic cells.
Recommendation
Supplement with 1.125 g/L of sodium bicarbonate for increased buffering capability and cell performance
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only