McCoy’s 5A Medium with L-Glutaminer, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
McCoy’s 5A Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. McCoy’s 5A Medium uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
- With L-glutamine and Phenol Red
McCoy’s 5A Medium originated from Dr. McCoy and his coworkers, as it was centered on the amino acid requirements of Novikoff hepatoma cells in culture. This work involved a medium known as Basal Medium 5A and used to grow normal bone marrow, skin, gingiva, testes, mouse kidney, omentum, adrenal glands, lung, spleen, rat embryos and other tissues. This product also includes Dr. Hsu’s addition of Hanks’ salts to enable use outside a CO2 incubator.
McCoy’s 5A Medium with L-glutamine will support growth of primary mammalian cells derived from the following: normal bone marrow skin, spleen, kidney, lung, rat embryos, and other tissues.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
A 10% FBS supplementation is recommended.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...