Medium 199 (E-199), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
112-022-101
10128-616EA 73.34 USD
10128-616
Medium 199 (E-199), Quality Biological
Cell Culture Media
E199 or Medium 199 with Earle’s Salt is one of the best known media in the history of cell culture. Morgan, Morton and Parker’s Medium 199 was the first really successful attempt to develop a totally defined medium. Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts.
  • With Phenol Red
  • Without L-glutamine, Hepes
E199 has been used for many different types of cells, especially non-transformed cells, and proved particularly useful in polio vaccine production and other virology applications such as vaccine production and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. QBI offers a variety of E-199 Medium modifications for a range of cell culture applications.
Sterile Filtered
Osmolality: 290±5%mOsm/Kg H20
Harmonization Code: 3821.00.0000
ECCN: EAR99
Because DMEM: F-12 has no lipids proteins and growth factors, DMEM: F-12 may require10% FBS supplementation. Requires 5-10% CO2 environment to maintain physiological pH.
Morgan, Morton and Parker's Medium 199 was the first really successful attempt to develop a totally defined medium. Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only