Ampicillin 100 mg/mL ready-to-use, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

351-344-731
10128-594PK 222.52 USD
10128-594
Ampicillin 100 mg/mL ready-to-use, sterile filtered
Ampicillin

Ampicillin in a convenient ready-to-use solution useful in selecting for ampicillin resistant bacteria. Ampicillin is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure.


  • Convenient 1000X concentrated stock
  • Small 1 mL size to prevent freeze-thaw cycle
  • Affordable price


Ampicillin, which is a semi-synthetic derivative of penicillin, inhibits cell wall synthesis (peptidoglycan crosslinking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane in Gram-negative and Gram-positive bacteria.2,3 Cleavage of the β-lactam ring of ampicillin by β-lactamase results in bacterial resistance to this antibiotic.

Product Contents
Ampicillin and Molecular Biology Grade Water

Recommendation
The recommended concentration for antibacterial use in culture media is ~100 mg/mL (a 1000-fold dilution of the product). The recommended concentration for use in ampicillin resistance studies is 20–125 mg/mL.

Functional Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: on dry ice


Caution: Research Use Only

Formula: C₁₆H₁₉N₃O₄S
Storage Temperature: Freezer
MDL Number: MFCD00005175
CAS Number: 69-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR