Customized SDS Protein Gel Loading Solution, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-151-681
10128-562PK 246.58 USD
10128-562 10128-564
Customized SDS Protein Gel Loading Solution, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers
This is a special formulation of loading solution for protein gel electrophoresis.
  • SDS-PAGE
  • Western Blotting
  • Analysis of DNA samples with high amounts of DNA binding proteins
  • Kinetic experiments
  • DNA agarose gel analysis after DNA restriction digestions, ligation, or dephosphorylation
Recommendation
For complex mixtures, use 50 µg total protein per lane.
For purified proteins, use 0.5-5 µg total protein per lane.
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.