Sodium dodecyl sulfate (SDS) 20% in aqueous solution, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

351-066-101 351-066-721 351-066-721EA
10128-504EA 138.29 USD
10128-504 10128-506 10128-508
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, sterile filtered
Sodium dodecyl sulphate (SDS)

Sodium Dodecyl Sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods; this SDS is lauryl-grade.


  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
  • Common component of cell lysis buffers
  • Filtered through a 0.22 µm membrane
  • Dnase, Rnase, and Protease tested


Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant.

Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR