TBS HCl (10X) Buffer Solution, pH 7.2, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-058-101
10128-500EA
65.65
USD
10128-500
TBS HCl (10X) Buffer Solution, pH 7.2, Quality Biological
Buffers
Electrophoresis Buffers
Washing Buffers
Tris HCl, 10X pH 7.2 is a Buffered Saline Solution used in procedures for the isolation of DNA.
- Filtered through a 0.22 µm membrane
- DNase, RNase, and Protease tested
Tris buffered saline is useful for the following: as a biological diluent buffer, as a buffer for antibodies, as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.