Potassium chloride 1 M in aqueous solution, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

351-044-101
10128-494EA 46.42 USD
10128-494
Potassium chloride 1 M in aqueous solution, sterile filtered
Potassium chloride

Potassium Chloride 1 M is used to establish a constant ionic strength in a sample solution.


  • KCl 74.56 g/L
  • Suitable for PCR, molecular biology, protein chemistry and biochemical applications
  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested


Potassium Chloride 1 M is used in molecular biology especially for DNA and RNA isolation procedures.

Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Appearance clear colorless liquid pass
Dnase, Rnase and Protease not detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR