Sodium chloride 5 M in aqueous solution, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

351-036-101 351-036-151 351-036-491 351-036-721 351-036-721EA
10128-484EA 41.73 USD
10128-484 10128-486 10128-488 10128-490 10128-492
Sodium chloride 5 M in aqueous solution, sterile filtered
Sodium chloride

Sodium Chloride, 5M is suitable for molecular biology, protein chemistry and biochemical applications.


  • 5 M concentration
  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested


Sodium Chloride, 5 M is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures.

UNSPSC: 41105514
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR