Magnesium chloride 2 M in aqueous solution, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

340-034-721 340-034-721EA
10128-296PK 223.41 USD
10128-296 10128-298
Magnesium chloride 2 M in aqueous solution, sterile filtered
Magnesium chloride

Magnesium is a typical ionic halide, and is very soluble in water. Magnesium has a variety of biological roles in enzymology, cell membrane/wall structure, muscle cell physiology, and nucleic acid structure. Magnesium is an essential co-factor in many enzymes, including DNAse, some restriction enzymes, and Ribonuclease H.


  • Used with SOC medium


Magnesium Chloride, 2 M can be added to SOB and SOC medium at a concentration of 20 mM (10 mL 2 M MgCl2 per liter).

UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR