Super Broth, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
Super Broth is used for the mass cultivation of Escherichia coli for high yield protein and plasmid DNA production.
- Has 220% more peptone and 300% more yeast extract than LB
Super Broth is the best commonly used E. Coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.
Sterile Filtered and
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Endotoxin and performance tested
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...