QBSF Serum Free Media, Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

160-204-101
10128-230EA 163.9 USD
10128-230 10128-232 10128-236
QBSF Serum Free Media, Quality Biological
Cell Culture Media

QBSF is a proprietary serum-free media line of four media. Each designed for specific applications where serum-free conditions are required, or more advantageous.


  • Serum-free
  • Contains L-glutamine and phenol red
  • Endotoxin, mycoplasma and performance tested
  • Sterile filtered (0.1 µm)
  • Osmolality: 285±3% mOsm/Kg H2O


QBSF-55 is a proprietary low protein (67.4±15% µg/ml) serum-free medium. It is designed to support the growth of hybridomas and enhance the production of monoclonal antibodies. The low protein concentration in QBSF-55 simplifies the downstream processing and recovery of monoclonal antibodies. Using this reduces target protein losses due to co-separation with serum or other proteins present in high concentration.


QBSF-56 is a proprietary serum-free formulation engineered to support lymphocytes blastogenesis. In addition, this can be used as an intermediate medium in the process of weaning fastidious cells from growth in a serum-supplemented medium to a low protein serum-free medium. It has also been used successfully to support the proliferation of primary cell cultures.


QBSF-58 is a proprietary serum-free medium designed to support murine and human bone marrow cell growth. This supports the differentiation of human or murine stem and/or progenitor cells to erythrocytes, granulocytes, and lymphocytes in the presence of the appropriate cytokines, It is favorable for both short-term and long-term expansion of marrow repopulating cells.


QBSF-60 is a proprietary serum-free medium that supports the expansion of human hematopoietic stem/progenitor cells. It supports CD34+ cells derived from normal bone marrow, cord blood, peripheral blood stem cells mobilized with cytokines for ex-vivo expansion and differentiation to erythrocytes, granulocytes, and lymphocytes.


The QBSF products have been used successfully used in different cell culture applications, investigators are encouraged to explore new uses.


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR