Kanamycin 50 mg/mL ready-to-use, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

120-349-731
10128-224PK 172.26 USD
10128-224
Kanamycin 50 mg/mL ready-to-use, sterile filtered
Kanamycin

Kanamycin sulfate is a water soluble broad spectrum antibiotic obtained from soil bacterium Streptomyces kanamyceticus. It is an aminoglycoside antibiotic effective against Gram-negative and Gram-positive bacteria and mycoplasma. It binds to the 30S ribosomal subunit, inhibits translocation, and elicits miscoding.


  • Convenient 1 mL vial ready to use.


Kanamycine ready-made 50 mg/mL solution acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding. Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Kanamycin is used in Molecular Biology as a selective agent most commonly to isolate bacteria which have taken up genes coupled to a gene coding for kanamycin resistance.

UNSPSC: 41100000
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research Use Only

MDL Number: MFCD00070289
CAS Number: 59-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR