DuPont™ Tyvek® 400 FC Shoe Covers with Skid Resistant Sole
Supplier: DuPont
FC450SGY00020000
10128-148CS 222.42 USD
10128-148 10016-034
DuPont™ Tyvek® 400 FC Shoe Covers with Skid Resistant Sole
Disposable Garments
Tyvek® 400 FC protective fabric features a skid resistant friction coating.
  • Skid-resistant sole
  • Elastic top
  • One size fits most
  • Serged seams
Used exclusively for shoe and boot covers and skid-resistant booties in select Tyvek® coveralls.
Certifications: Thickness meets ASTM D1777.
Ordering information: Available standard or NAFTA sourced.