TSHP Series FMS High Performance Laboratory Refrigerators, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSFMS2305A
10127-732EA 12374.5 USD
10127-732
TSHP Series FMS High Performance Laboratory Refrigerators, Thermo Scientific
Refrigerators
These refrigerators maintain a safe environment while keeping valuable samples within a tight temperature range.
  • Intuitive digital display featuring microprocessor control
  • Convenient access port and casters
  • Lockable doors, audio and visual alarms including door ajar alarms for security
Designed to meet stringent U.S. Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable materials, solvents and intermediates.
Outstanding construction featuring painted, high-quality steel exteriors with durable finish.Manual defrost and ignition-free interior cabinet design for the safe storage of flammable materials.
Certifications: UL listed.
Ordering information: Temperature chart recorders, and 4 to 20 milliamp are available separately.