Revco® FMS High-Performance Laboratory Refrigerators, Thermo Scientific
Supplier: Thermo Fisher Scientific
RFMS2305A
10127-730EA 13999.86 USD
10127-730
Revco® FMS High-Performance Laboratory Refrigerators, Thermo Scientific
Refrigerators
These refrigerators are designed to maintain a safe environment while keeping valuable samples within a tight temperature range.
  • Intuitive digital display featuring microprocessor control
  • Convenient access port and casters
  • Lockable doors, audio and visual alarms including door ajar alarms for security
Designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids, solvents and intermediates.
Outstanding construction featuring painted, high-quality steel exteriors with durable finish.Manual defrost and ignition-free interior cabinet design for the safe storage of flammable materials.
Certifications: UL listed.
Ordering information: Temperature chart recorders, and 4-20 milliamp are available separately.