ULTIMATE 70™ Sealed-Edge Polyester Knit Heavy Weight Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
ULT70-99
10127-662BG 77.44 USD
10127-662
ULTIMATE 70™ Sealed-Edge Polyester Knit Heavy Weight Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ultimate 70™ laser sealed-edge heavyweight wipes are extremely absorbent, abrasion resistant, and low in particle emission.
  • Polyester knit
  • Sealed-edge
  • Laundered
  • Cleanroom packaging
Ultimate 70™ are a heavyweight, 100% polyester cleanroom wipe with laser sealed edges. The heat of the laser beam cuts and melts each fiber along the edges, reducing the chances of particle emission. Each wipe is laundered in a class 10 cleanroom, individually inspected, and sealed in cleanroom bags. These wipes are extremely absorbent, abrasion resistant, and low in ionic levels.
Packaging: Packaged 150 wipes per bag, 10 bags per case.