Latitude® Midigel, 2%

Supplier: Lonza
57239
10127-418PK 430.17 USD
10127-418
Latitude® Midigel, 2%
Electrophoresis Gels Precast Gels
Precast Latitude® midigels TBE are made for accurate DNA fragment separation for most any gel electrophoresis application. They are manufactured from high quality, Lonza molecular biology grade SeaKem® LE Plus Agarose.

  • Fast - No more pouring or cooling times, easy - simply unwrap and use, traceable - trackable lots

These ready-to-run gels are available in two different well formats and agarose concentrations in TBE buffer to fit most DNA electrophoresis needs. Our Latitude® Precast Agarose Gels are precision cast for accurate, reproducible resolution of DNA fragments for 100 bp to 8 kb and pre stained with 0.5 µg/ml Ethidium Bromide. Latitude® Midigels TBE fit most standard electrophoresis chambers and are stable for 6 to 12 months, depending on their concentration.

Specifications:
Separation range 100 bp to 2 kb
Gel dimensions 10×15 cm
Gel thickness 6.0 mm
Ethidium bromide 0.5 μg/ml
Tray dimensions 10.4×15.6 cm
Well volume10 to 12 μl

Applications: DNA Electrophoresis.

Delivery information: Content: Latitude® Midigel (10×15 cm) 2% SeaKem® LE Agarose, 15-well, 8 gels.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR