Bucket, AlphaMop™ TX7054, Texwipe
Supplier: Texwipe
TX7054
10127-094EA 723.01 USD
10127-094
Bucket, AlphaMop™ TX7054, Texwipe
Buckets
Made from 100% 304 stainless steel.
  • Perfect for cleaning walls, ceilings and floors
  • Ideal for use with common disinfectants and cleaners, including Texwipe’s TexQ
  • Can be used in ISO Class 3 - 7 environments
Bucket physical dimensions: 21"L x 13"W x 8"H (53cm x 33cm w x 20cm) with a 7-gallon (28 liter) capacity.