Mega Tube Racks for 10 - 13 mm Tubes
Supplier: Heathrow Scientific
HS24322A HS24322E
10124-742CS 229.12 USD
10124-742 10124-790
Mega Tube Racks for 10 - 13 mm Tubes
Racks Tube Racks
These fully assembled, mega tube racks are ideal for high throughput labs requiring large sample storage.
  • Sturdy and lightweight material for easy and secure transport of blood collection tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate the need for multiple tube racks
  • Racks are manufactured from polypropylene and are autoclavable
  • High-capacity storage for 5 to 7 ml tubes