Phenol Red Broth with Dextrose, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Y304
10124-528PK 211.61 USD
10124-528
Phenol Red Broth with Dextrose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of microorganisms by carbohydrate fermentation.
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.