Phenol Red Broth with Lactose, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Y309
10124-526PK 337.53 USD
10124-526
Phenol Red Broth with Lactose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of microorganisms by carbohydrate fermentation.
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.