Phosphate Buffer
Supplier: HARDY DIAGNOSTICS
U438
10124-486PK 361.59 USD
10124-486
Phosphate Buffer
Media for Microbiology Dehydrated Media for Microbiology
Recommended for use as a diluent in the preparation of dilutions for plate counts and other laboratory processes for non-clinical samples.
Recommended for use as a diluent in the preparation of dilutions for plate counts and other laboratory processes for non-clinical samples. Not intended for use in the diagnosis of human disease.