VWR® Cryogenic Dot and Strip Labels
Supplier: VWR International
89097-572 89097-574 89097-576 89097-578 89097-580 89097-582 89097-584 89097-586 89097-590 89097-592 89097-594 89097-596 89097-598 89097-600 89097-602 89097-604 89097-606 89097-608 89097-610 89097-612 89097-614 89097-616 89129-280 89129-282 89129-284 89129-286 89129-288 89129-290 89129-292 89129-294 89097-588 10123-758 10123-760 10123-762 10123-764 10123-766 10123-768 10123-770 10123-772 10123-774 10123-776 10123-778 10123-780 10123-782 10123-784 10123-786 10123-788 10123-790 10123-792 10123-794 10123-796 10123-798 10123-800 10123-802 10123-804 10123-806 10123-808 10123-810 10123-812 10123-814 10123-816 10123-818 10123-820 10123-822 10123-824 10123-826 10123-828 10123-830 10123-832 10123-834 10123-836 10123-838 10123-840 10123-842 10123-844 10123-846 10123-848 10123-852 10123-854 10123-856 10123-858 10123-860 10123-862 10123-864 10123-866 10123-868 10123-870 10123-872 10123-874
89097-572CS 51.68 USD
89097-572 89097-574 89097-576 89097-578 89097-580 89097-582 89097-584 89097-586 89097-590 89097-592 89097-594 89097-596 89097-598 89097-600 89097-602 89097-604 89097-606 89097-608 89097-610 89097-612 89097-614 89097-616 89129-280 89129-282 89129-284 89129-286 89129-288 89129-290 89129-292 89129-294 89097-588 10123-758 10123-760 10123-762 10123-764 10123-766 10123-768 10123-770 10123-772 10123-774 10123-776 10123-778 10123-780 10123-782 10123-784 10123-786 10123-788 10123-790 10123-792 10123-794 10123-796 10123-798 10123-800 10123-802 10123-804 10123-806 10123-808 10123-810 10123-812 10123-814 10123-816 10123-818 10123-820 10123-822 10123-824 10123-826 10123-828 10123-830 10123-832 10123-834 10123-836 10123-838 10123-840 10123-842 10123-844 10123-846 10123-848 10123-852 10123-854 10123-856 10123-858 10123-860 10123-862 10123-864 10123-866 10123-868 10123-870 10123-872 10123-874
VWR® Cryogenic Dot and Strip Labels
Labels
VWR® Cryogenic Dot and Strip Labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Labels can be placed in most incubators
Labels feature a unique adhesive that can withstand extreme temperatures from –196...121 °C (–321...250 °F), without the need for lamination.
Ordering information: Rolls of labels are supplied on a 44 mm Ø (1³/₄") core. Laser printer label sheets are 21.6W×27.9L cm (8¹/₂×11"). Assorted labels come in blue, green, purple, red, and yellow. Roll of assorted labels contains 1000 labels of each color. Boxes of assorted laser printer labels contain five sheets of each color.