Random Primer 9, New England Biolabs
Supplier: New England Biolabs (NEB)
S1254S
101229-750EA 119 USD
101229-750
Random Primer 9, New England Biolabs
Nucleic Acid Reagents Primers and Probes
This mixture of random nonanucleotides is used to prime DNA synthesis in vitro along multiple sites of denatured template DNA
The newly synthesized complementary DNA is "oligo-labelled" by substituting any radiolabelled nucleotide for the appropriate non-radioactive nucleotide in the reaction mixture. Use of synthetic d(N9) primer ensures the presence of virtually all sequence combination of nanomer primers which results in equally labelled DNA of high specific activity. Oligolabelling by this method generates probes which can be used to screen gene libraries, probe Southern and Northern blots, and for in situ hybridizations.
This primer-template complex is a suitable substrate for DNA Polymerase I(Klenow Fragment).