Acyclonucleotide Set, New England Biolabs
Supplier: New England Biolabs (NEB)
N0460S
101228-420EA 85 USD
101228-420
Acyclonucleotide Set, New England Biolabs
Nucleic Acid Reagents Nucleotides
Acyclonucleotide Set contains four separate tubes of acyNTPs (acyATP, acyCTP, acyGTP and acyTTP).
Acyclonucleotides are supplied as a dry powder. Addition of 50 µl of distilled or de-ionized (Milli-Q®) water will result in a  final concentration of 10 mM acyNTP. Acyclonucleotides (acyNTPs) act as chain terminators and are thus useful in applications that normally employ dideoxynucleotides such as DNA sequencing and SNP detection. AcyNTPs are especially useful in applications with archaeon DNA Polymerases, more specifically with Therminator™ DNA Polymerase. Therminator DNA Polymerase is an engineered enzyme with an increased capacity to incorporate analogs with altered sugars, such as ribonucleotides, dideoxynucleotides, 2´ deoxynucleotides and especially acyclo-base analogs.