Deoxynucleotide Solution Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
Deoxynucleotide (dNTP) Solution Mix is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP.
Each vial contains 8 μmol in a volume of 0.8 ml (10 mM concentration). A large pack contains 5 vials of 0.8 ml.
Delivery information: Supplied in ultrapure water as a sodium salt at pH 7.5.