ssRNA Ladder, New England Biolabs
Supplier: New England Biolabs (NEB)
N0362S
101228-410EA 85 USD
101228-410
ssRNA Ladder, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The ssRNA Ladder is a set of 7 RNA molecules produced by in vitro transcription of a mixture of 7 linear DNA templates
The ladder sizes are: 9000, 7000, 5000, 3000, 2000, 1000 and 500 bases. The 3000 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing or native agarose gels.