T4 DNA Ligase, New England Biolabs
Supplier: New England Biolabs (NEB)
Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA.
This enzyme will join blunt end and cohesive end termini as well as repair single stranded nicks in duplex DNA and some DNA/RNA hybrids. T4 DNA ligase will seal nicks for these DNA substrates.