Chitin Magnetic Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
E8036S
101227-556EA 152 USD
101227-556
Chitin Magnetic Beads, New England Biolabs
Beads Magnetic Beads
An affinity matrix for the small-scale isolation of target proteins fused to a chitin binding domain (CBD)
Chitin beads have been prepared having a magnetite core. This permits the magnetic isolation of CBD-fusion proteins from cell culture supernatants; after which, the matrix can be regenerated without loss of binding capacity. Immobilized fusion proteins can be used in subsequent experiments to capture target proteins from crude cell lysates that interact with the immobilized fusion protein.