Standard Taq (Mg Free) Reaction Buffer Pack, New England Biolabs
Supplier: New England Biolabs (NEB)
B9015S
101227-508EA 27 USD
101227-508
Standard Taq (Mg Free) Reaction Buffer Pack, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
New England Biolabs supplies a unique 10X reaction buffer and a supplemental reagent for use with Taq DNA Polymerase in reaction systems which require a detergent free, low ionic strength buffer and variable magnesium concentration.