THERMOPOL® Reaction Buffer Pack , New England Biolabs
Supplier: New England Biolabs (NEB)
B9004S
101227-498EA 27 USD
101227-498
THERMOPOL® Reaction Buffer Pack , New England Biolabs
Buffers Cloning Buffers
10X ThermoPol Reaction Buffer is optimized for use with VentR® and Deep VentR™ DNA Polymerases
This buffer also provides superior reaction conditions for other thermophilic DNA polymerases, including Taq DNA Polymerase, as well as various other DNA and RNA modifying enzymes. Supplemental MgSO4 is provided to maximize flexibility for users that require additional Mg2+ for their specific application.