Deoxynucleotide Solution Set, New England Biolabs
Supplier: New England Biolabs (NEB)
N0446S
101227-334EA 206 USD
101227-334
Deoxynucleotide Solution Set, New England Biolabs
Nucleic Acid Reagents Nucleotides
The Deoxynucleotide (dNTP) Solution Set contains 0.25 ml of each solution of ultrapure deoxynucleotide (dATP, dCTP, dGTP and dTTP)
Each deoxynucleotide is supplied at a concentration of 100 mM. Supplied in: Ultrapure water as a sodium salt at pH 7.5.