Oligo d(T)18 (no 5'-Phosphate), New England Biolabs
Supplier: New England Biolabs (NEB)
S1316S
101227-236EA 111 USD
101227-236
Oligo d(T)18 (no 5'-Phosphate), New England Biolabs
Nucleic Acid Reagents Primers and Probes
This primer is used for the priming and sequencing of mRNA adjacent to the 3'-poly A tail or tailed cDNA
This primer does not contain a 5'-phosphate.