TRIDYE™ DNA Ladder, 100 bp , New England Biolabs
Supplier: New England Biolabs (NEB)
N3271S
101227-144EA 109 USD
101227-144
TRIDYE™ DNA Ladder, 100 bp , New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
TriDye™ 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis
The DNA ladder consists of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference points.