TRIDYE™ DNA Ladder, 1 kb Plus, New England Biolabs
Supplier: New England Biolabs (NEB)
N3270S
101227-142EA 99 USD
101227-142
TRIDYE™ DNA Ladder, 1 kb Plus, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
TriDye™ 2-Log DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference points.