Sulfuric acid ≥93%, Technical Grade

Supplier: AQUA SOLUTIONS MS

S9243-4L S9243-20L
101226-670EA 78.19 USD
101226-670 101225-652
Sulfuric acid ≥93%, Technical Grade
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR