Silver nitrate 0.001 N

Supplier: AQUA SOLUTIONS MS

8381-1L
101226-464EA 71.66 USD
101226-464
Silver nitrate 0.001 N
Silver nitrate
Formula: AgNO₃
MDL Number: MFCD00003414
CAS Number: 7761-88-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR